missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-beta,beta-difluorostyrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. B28561G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28561G Supplier TCI America Supplier No. B28561G
Only null left

Chemical Identifiers

CAS 84750-93-6
Molecular Formula C8H5BrF2
Molecular Weight (g/mol) 219.029
InChI Key LXJISEJYMAHRKE-UHFFFAOYSA-N
Synonym 1-Bromo-4-(2,2-difluorovinyl)benzene, 2-(4-Bromophenyl)-1,1-difluoroethylene
PubChem CID 22913822
IUPAC Name 1-bromo-4-(2,2-difluoroethenyl)benzene
SMILES C1=CC(=CC=C1C=C(F)F)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 219.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-beta,beta-difluorostyrene (stabilized with TBC)
TSCA No
Recommended Storage Freezer