accessibility menu, dialog, popup

UserName

CAS RN 84750-93-6

FFBr

CAS RN 84750-93-6

IUPAC Name: 1-bromo-4-(2,2-difluoroethenyl)benzene
Synonyms: 1-bromo-4-(2,2-difluoroethenyl)benzene
Molecular Weight (g/mol): 219.03
Molecular Formula: C8H5BrF2
InChi Key: LXJISEJYMAHRKE-UHFFFAOYSA-N
SMILES: FC(F)=CC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results