missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-tert-butylbiphenyl, 98%

Catalog No. AAH2707603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2707603 1 g
AAH2707609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2707603 Supplier Thermo Scientific Chemicals Supplier No. H2707603

CAS: 162258-89-1 | C16H17Br | 289.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162258-89-1
Molecular Formula C16H17Br
Molecular Weight (g/mol) 289.22
MDL Number MFCD01321141
InChI Key QYNWFBYWVPMMRL-UHFFFAOYSA-N
Synonym 4-bromo-4'-tert-butylbiphenyl, 1,1'-biphenyl,4-bromo-4'-1,1-dimethylethyl, 4-bromo-4'-tert-butyl-1,1'-biphenyl, acmc-209do3, 4-bromo-4'-tertbutylbiphenyl, 4'-bromo-4-tert-butyl-biphenyl, 4-bromo-4/'-tert-butylbiphenyl, 4-bromo-4-tert-butylbiphenyl, 1-4-bromophenyl-4-tert-butylbenzene
PubChem CID 9971389
SMILES CC(C)(C)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 140°C to 143°C
Boiling Point 152°C (2 mmHg)
Quantity 1 g
IUPAC Name 4-bromo-4'-tert-butyl-1,1'-biphenyl
Formula Weight 289.21
Percent Purity 98%
Chemical Name or Material 4-Bromo-4'-tert-butylbiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only