accessibility menu, dialog, popup

UserName

CH3H3CCH3Br

CAS RN 162258-89-1

IUPAC Name: 4-bromo-4'-tert-butyl-1,1'-biphenyl
Synonyms: 4-bromo-4'-tert-butyl-1,1'-biphenyl
Molecular Weight (g/mol): 289.22
Molecular Formula: C16H17Br
InChi Key: QYNWFBYWVPMMRL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)