missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-propylbiphenyl 98.0+%, TCI America™

Catalog No. B13065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13065G 5 g
B130625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13065G Supplier TCI America Supplier No. B13065G
Only null left

Chemical Identifiers

CAS 58743-81-0
Molecular Formula C15H15Br
Molecular Weight (g/mol) 275.19
MDL Number MFCD00060105
InChI Key DPERLOMMOVDOHH-UHFFFAOYSA-N
PubChem CID 619019
IUPAC Name 4-bromo-4'-propyl-1,1'-biphenyl
SMILES CCCC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 5 g
Formula Weight 275.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-propylbiphenyl