accessibility menu, dialog, popup

UserName

CH3Br

CAS RN 58743-81-0

IUPAC Name: 4-bromo-4'-propyl-1,1'-biphenyl
Synonyms: 4-bromo-4'-propyl-1,1'-biphenyl
Molecular Weight (g/mol): 275.19
Molecular Formula: C15H15Br
InChi Key: DPERLOMMOVDOHH-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results