missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-4'-hydroxybiphenyl 99.0+%, TCI America™
Supplier: TCI America H06905G
Specifications
| 4-Bromo-4′-hydroxybiphenyl | |
| 168°C | |
| C12H9BrO | |
| 5 g | |
| ARUBXNBYMCVENE-UHFFFAOYSA-N | |
| 4-(4-bromophenyl)phenol | |
| 95093 | |
| ≥99.0% (GC) |
| 29558-77-8 | |
| White | |
| MFCD00059076 | |
| 4-bromo-4'-hydroxybiphenyl, 4'-bromo-1,1'-biphenyl-4-ol, 4-4-bromophenyl phenol, 4'-bromobiphenyl-4-ol, 4-hydroxy-4'-bromobiphenyl, 1,1'-biphenyl-4-ol, 4'-bromo, 4-bromo-4-hydroxybiphenyl, pubchem23025 | |
| C1=CC(=CC=C1C2=CC=C(C=C2)Br)O | |
| 249.107 | |
| 249.11 | |
| Crystalline Powder |
Chemical Identifiers
| 29558-77-8 | |
| 249.107 | |
| ARUBXNBYMCVENE-UHFFFAOYSA-N | |
| 95093 | |
| C1=CC(=CC=C1C2=CC=C(C=C2)Br)O |
| C12H9BrO | |
| MFCD00059076 | |
| 4-bromo-4'-hydroxybiphenyl, 4'-bromo-1,1'-biphenyl-4-ol, 4-4-bromophenyl phenol, 4'-bromobiphenyl-4-ol, 4-hydroxy-4'-bromobiphenyl, 1,1'-biphenyl-4-ol, 4'-bromo, 4-bromo-4-hydroxybiphenyl, pubchem23025 | |
| 4-(4-bromophenyl)phenol |
Safety and Handling
TSCA : No