missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-hydroxybiphenyl 99.0+%, TCI America™

Catalog No. p-7130674
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06905G 5 g
H069025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06905G Supplier TCI America Supplier No. H06905G
Only null left

Chemical Identifiers

CAS 29558-77-8
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00059076
InChI Key ARUBXNBYMCVENE-UHFFFAOYSA-N
Synonym 4-bromo-4'-hydroxybiphenyl, 4'-bromo-1,1'-biphenyl-4-ol, 4-4-bromophenyl phenol, 4'-bromobiphenyl-4-ol, 4-hydroxy-4'-bromobiphenyl, 1,1'-biphenyl-4-ol, 4'-bromo, 4-bromo-4-hydroxybiphenyl, pubchem23025
PubChem CID 95093
IUPAC Name 4-(4-bromophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Br)O

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 249.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-hydroxybiphenyl