missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-(trifluoromethyl)phenol 98.0+%, TCI America™

Catalog No. B44941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44941G 1 g
B44945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44941G Supplier TCI America Supplier No. B44941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 320-49-0
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD03095342
InChI Key VOWPIDJSINRFPZ-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl phenol, 2-bromo-5-hydroxybenzotrifluoride, 3-trifluoromethyl-4-bromophenol, phenol, 4-bromo-3-trifluoromethyl, 4-bromo-3-trifluromethylphenol, 4-bromo-3-trifluoromethyl-phenol, btfp, pubchem2111, acmc-1ahmz, 3-trifluoromethyl-4-bromo phenol
PubChem CID 22277307
IUPAC Name 4-bromo-3-(trifluoromethyl)phenol
SMILES OC1=CC(=C(Br)C=C1)C(F)(F)F

Specifications

Melting Point 48°C
Quantity 1 g
Formula Weight 241.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)phenol
TSCA No
Recommended Storage Refrigerator