accessibility menu, dialog, popup

UserName

CAS RN 320-49-0

IUPAC Name: 4-bromo-3-(trifluoromethyl)phenol
Synonyms: 4-bromo-3-(trifluoromethyl)phenol
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: VOWPIDJSINRFPZ-UHFFFAOYSA-N
SMILES: OC1=CC(=C(Br)C=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results