missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. B46915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46915G 5 g
B46911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46915G Supplier TCI America Supplier No. B46915G

Chemical Identifiers

CAS 1735-53-1
Molecular Formula C8H3BrF3N
Molecular Weight (g/mol) 250.018
MDL Number MFCD03095347
InChI Key KSXUIQQDHHFSRN-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl benzonitrile, 3-trifluoromethyl-4-bromobenzonitrile, 2-bromo-5-cyanobenzotrifluoride, 3-trifluoromethyl-4-bromo benzonitrile, benzonitrile, 4-bromo-3-trifluoromethyl, 4-bromo-3-trifluoromethyl benzenecarbonitrile, 4-bromo-3-trifluoromethyl-benzonitrile, pubchem1576, intermediates-zcf02288, acmc-209e6z
PubChem CID 10800647
IUPAC Name 4-bromo-3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C#N)C(F)(F)F)Br

Specifications

Melting Point 82°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 250.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)benzonitrile