accessibility menu, dialog, popup

UserName

NFFFBr

CAS RN 1735-53-1

IUPAC Name: 4-bromo-3-(trifluoromethyl)benzonitrile
Synonyms: 4-bromo-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 250.02
Molecular Formula: C8H3BrF3N
InChi Key: KSXUIQQDHHFSRN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Br)C=CC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results