missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrobenzaldehyde 98.0+%, TCI America™

$46.60 - $137.94

Chemical Identifiers

CAS 163596-75-6
Molecular Formula C7H4BrNO3
Molecular Weight (g/mol) 230.017
MDL Number MFCD00204031
InChI Key SAFSVELFSYQXOV-UHFFFAOYSA-N
PubChem CID 808792
IUPAC Name 4-bromo-3-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B51011G
View Documents
TCI America
B51011G
1 g
Each for $46.60
Only null left
 
B51015G
View Documents
TCI America
B51015G
5 g
Each for $137.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

163596-75-6
230.017
SAFSVELFSYQXOV-UHFFFAOYSA-N
4-bromo-3-nitrobenzaldehyde
C7H4BrNO3
MFCD00204031
808792
C1=CC(=C(C=C1C=O)[N+](=O)[O-])Br

Specifications

163596-75-6
C7H4BrNO3
1 g
C1=CC(=C(C=C1C=O)[N+](=O)[O-])Br
230.017
230.02
Crystalline Powder
105°C
MFCD00204031
SAFSVELFSYQXOV-UHFFFAOYSA-N
4-bromo-3-nitrobenzaldehyde
808792
≥98.0% (GC)
4-Bromo-3-nitrobenzaldehyde
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'