missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-nitrobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America B51015G
Specifications
| 4-Bromo-3-nitrobenzaldehyde | |
| 105°C | |
| MFCD00204031 | |
| SAFSVELFSYQXOV-UHFFFAOYSA-N | |
| 4-bromo-3-nitrobenzaldehyde | |
| 808792 | |
| ≥98.0% (GC) |
| 163596-75-6 | |
| C7H4BrNO3 | |
| 5 g | |
| C1=CC(=C(C=C1C=O)[N+](=O)[O-])Br | |
| 230.017 | |
| 230.02 | |
| Crystalline Powder |
Chemical Identifiers
| 163596-75-6 | |
| 230.017 | |
| SAFSVELFSYQXOV-UHFFFAOYSA-N | |
| 4-bromo-3-nitrobenzaldehyde |
| C7H4BrNO3 | |
| MFCD00204031 | |
| 808792 | |
| C1=CC(=C(C=C1C=O)[N+](=O)[O-])Br |
Safety and Handling
TSCA : No