missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-nitroanisole 96.0+%, TCI America™
Supplier: TCI America B342025G
Specifications
| 4-Bromo-3-nitroanisole | |
| 32°C | |
| 154°C | |
| MFCD00051511 | |
| 4-bromo-3-nitroanisole, 4-bromo-3-nitroanisol, benzene, 1-bromo-4-methoxy-2-nitro, 3-nitro-4-bromoanisole, 4-methoxy-2-nitrobromobenzene, 4-bromo-3-nitrothioanisole, 4-bromo-3-nitrophenyl methyl ether, 1-bromo-4-methoxy-2-nitro-benzene, timtec-bb sbb009974, pubchem2655 | |
| COC1=CC=C(Br)C(=C1)[N+]([O-])=O | |
| 232.03 | |
| 232.03 | |
| Crystalline Lumps |
| 5344-78-5 | |
| White-Yellow | |
| C7H6BrNO3 | |
| 25 g | |
| KCOBIBRGPCFIGF-UHFFFAOYSA-N | |
| 1-bromo-4-methoxy-2-nitrobenzene | |
| 79288 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 5344-78-5 | |
| 232.03 | |
| KCOBIBRGPCFIGF-UHFFFAOYSA-N | |
| 79288 | |
| COC1=CC=C(Br)C(=C1)[N+]([O-])=O |
| C7H6BrNO3 | |
| MFCD00051511 | |
| 4-bromo-3-nitroanisole, 4-bromo-3-nitroanisol, benzene, 1-bromo-4-methoxy-2-nitro, 3-nitro-4-bromoanisole, 4-methoxy-2-nitrobromobenzene, 4-bromo-3-nitrothioanisole, 4-bromo-3-nitrophenyl methyl ether, 1-bromo-4-methoxy-2-nitro-benzene, timtec-bb sbb009974, pubchem2655 | |
| 1-bromo-4-methoxy-2-nitrobenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator