missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitroanisole 96.0+%, TCI America™

Catalog No. p-7122937
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34205G 5 g
B342025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34205G Supplier TCI America Supplier No. B34205G
Only null left

Chemical Identifiers

CAS 5344-78-5
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD00051511
InChI Key KCOBIBRGPCFIGF-UHFFFAOYSA-N
Synonym 4-bromo-3-nitroanisole, 4-bromo-3-nitroanisol, benzene, 1-bromo-4-methoxy-2-nitro, 3-nitro-4-bromoanisole, 4-methoxy-2-nitrobromobenzene, 4-bromo-3-nitrothioanisole, 4-bromo-3-nitrophenyl methyl ether, 1-bromo-4-methoxy-2-nitro-benzene, timtec-bb sbb009974, pubchem2655
PubChem CID 79288
IUPAC Name 1-bromo-4-methoxy-2-nitrobenzene
SMILES COC1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 154°C
Quantity 5 g
Formula Weight 232.03
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-3-nitroanisole
TSCA No
Recommended Storage Refrigerator