missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitroaniline 98.0+%, TCI America™

Catalog No. p-7123808
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48761G 1 g
B48765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48761G Supplier TCI America Supplier No. B48761G
Only null left

Chemical Identifiers

CAS 53324-38-2
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD01320678
InChI Key PITHQPMZWKZGRZ-UHFFFAOYSA-N
PubChem CID 612299
IUPAC Name 4-bromo-3-nitroaniline
SMILES NC1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 129°C
Quantity 1 g
Synonym 4-bromo-3-nitro-phenylamine, benzenamine, 4-bromo-3-nitro, 4-bromo-3-nitro-aniline, 4-bromo-3-nitrophenylamine, pubchem2613, 3-nitro-4-bromoaniline, 4-bromo-3-nitrobenzenamine, 4-bromanyl-3-nitro-aniline, ksc494a6p, benzenamine,4-bromo-3-nitro
Formula Weight 217.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-nitroaniline