accessibility menu, dialog, popup

UserName

CAS RN 53324-38-2

IUPAC Name: 4-bromo-3-nitroaniline
Synonyms: 4-bromo-3-nitroaniline
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: PITHQPMZWKZGRZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)