missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylpyrazole 98.0+%, TCI America™

$42.36 - $55.84

Chemical Identifiers

CAS 13808-64-5
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.002
MDL Number MFCD00005241
InChI Key IXQPRETWBGVNPJ-UHFFFAOYSA-N
Synonym 4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4
PubChem CID 83741
IUPAC Name 4-bromo-5-methyl-1H-pyrazole
SMILES CC1=C(C=NN1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B35221G
View Documents
TCI America
B35221G
1 g
Each for $42.36
Only null left
 
B35225G
View Documents
TCI America
B35225G
5 g
Each for $55.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13808-64-5
161.002
IXQPRETWBGVNPJ-UHFFFAOYSA-N
83741
CC1=C(C=NN1)Br
C4H5BrN2
MFCD00005241
4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4
4-bromo-5-methyl-1H-pyrazole

Specifications

13808-64-5
White-Yellow
MFCD00005241
4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4
CC1=C(C=NN1)Br
161.002
161.00
Crystalline Powder
76°C
C4H5BrN2
1 g
IXQPRETWBGVNPJ-UHFFFAOYSA-N
4-bromo-5-methyl-1H-pyrazole
83741
≥98.0% (GC)
4-Bromo-3-methylpyrazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents