missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-methylpyrazole 98.0+%, TCI America™
Supplier: TCI America B35225G
Specifications
| 4-Bromo-3-methylpyrazole | |
| 76°C | |
| C4H5BrN2 | |
| 5 g | |
| IXQPRETWBGVNPJ-UHFFFAOYSA-N | |
| 4-bromo-5-methyl-1H-pyrazole | |
| 83741 | |
| ≥98.0% (GC) |
| 13808-64-5 | |
| White-Yellow | |
| MFCD00005241 | |
| 4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4 | |
| CC1=C(C=NN1)Br | |
| 161.002 | |
| 161.00 | |
| Crystalline Powder |
Chemical Identifiers
| 13808-64-5 | |
| 161.002 | |
| IXQPRETWBGVNPJ-UHFFFAOYSA-N | |
| 83741 | |
| CC1=C(C=NN1)Br |
| C4H5BrN2 | |
| MFCD00005241 | |
| 4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4 | |
| 4-bromo-5-methyl-1H-pyrazole |
Safety and Handling
TSCA : No