accessibility menu, dialog, popup

UserName

CAS RN 13808-64-5

IUPAC Name: 4-bromo-5-methyl-1H-pyrazole
Synonyms: 4-bromo-3-methyl-2H-pyrazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: IXQPRETWBGVNPJ-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=NN1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)