Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-methylphenol, 98%

Catalog No. p-2690600
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368200050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC368200050 Supplier Thermo Scientific Chemicals Supplier No. 368200050

CAS: 14472-14-1 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14472-14-1
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00079723
InChI Key GPOQODYGMUTOQL-UHFFFAOYSA-N
Synonym 4-bromo-3-methyl-phenol, 4-bromo-m-cresol, phenol, 4-bromo-3-methyl, 4-bromo-3-methyl phenol, 4-bromo-5-methyl-phenol, 4-brom-3-methylphenol, zlchem 434, pubchem2201, 3-methyl-4-bromophenol, 2-bromo-5-hydroxytoluene
PubChem CID 72857
IUPAC Name 4-bromo-3-methylphenol
SMILES CC1=CC(O)=CC=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 57°C to 61°C
Color White
Boiling Point 142°C to 145°C (23.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(CH3)OH
Quantity 5 g
Beilstein 06, IV, 1321
Formula Weight 187.04
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Bromo-3-methylphenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-464-3

RUO – Research Use Only