missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. B52855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52855G 5 g
B52851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52855G Supplier TCI America Supplier No. B52855G

Chemical Identifiers

CAS 149104-89-2
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD09264005
InChI Key HBSHHYYUASJVCG-UHFFFAOYSA-N
PubChem CID 10987288
IUPAC Name (4-bromo-3-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)CO)Br

Specifications

Melting Point 36°C
Boiling Point 90°C
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylbenzyl Alcohol
TSCA No
Recommended Storage Refrigerator