accessibility menu, dialog, popup

UserName

CH3HOBr

CAS RN 149104-89-2

IUPAC Name: (4-bromo-3-methylphenyl)methanol
Synonyms: (4-bromo-3-methylphenyl)methanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: HBSHHYYUASJVCG-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(CO)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results