missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzoic acid, 98%

Catalog No. AAA1430214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1430214 25 g
AAA1430206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1430214 Supplier Thermo Scientific Chemicals Supplier No. A1430214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 7697-28-1 | C8H6BrO2 | 214.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7697-28-1
Molecular Formula C8H6BrO2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00039526
InChI Key KWVXDZLVCISXIB-UHFFFAOYSA-M
Synonym 4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b
PubChem CID 82131
IUPAC Name 4-bromo-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Br)C([O-])=O

Specifications

Melting Point 215°C to 218°C
Quantity 25 g
Beilstein 2356649
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-713-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only