missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-hydroxybenzyl alcohol, 95%, Thermo Scientific Chemicals

Catalog No. p-7056990
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66253MD 250 mg
AAH6625303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66253MD Supplier Thermo Scientific Chemicals Supplier No. H66253MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2737-19-1
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.04
MDL Number MFCD14705103
InChI Key JYYCVESONPFMEW-UHFFFAOYSA-N
PubChem CID 13551344
IUPAC Name 2-bromo-5-(hydroxymethyl)phenol
SMILES OCC1=CC(O)=C(Br)C=C1

Specifications

Quantity 250 mg
Beilstein 1936254
Synonym 2-bromo-5-hydroxymethyl phenol, 4-bromo-3-hydroxybenzyl alcohol, benzenemethanol, 4-bromo-3-hydroxy, 4-bromo-3-hydroxybenzyloxamine, 2-bromo-5-hydroxymethyl-phenol, 4-bromo-3-hydroxy-benzyl alcohol, benzenemethanol,4-bromo-3-hydroxy
Formula Weight 203.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-3-hydroxybenzyl alcohol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only