accessibility menu, dialog, popup

UserName

CAS RN 2737-19-1

IUPAC Name: 2-bromo-5-(hydroxymethyl)phenol
Synonyms: 2-bromo-5-(hydroxymethyl)phenol
Molecular Weight (g/mol): 203.04
Molecular Formula: C7H7BrO2
InChi Key: JYYCVESONPFMEW-UHFFFAOYSA-N
SMILES: OCC1=CC(O)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results