missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-hydroxy-2-naphthoic Acid 97.0+%, TCI America™

Catalog No. p-7122821
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32295G 5 g
B322925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32295G Supplier TCI America Supplier No. B32295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2208-15-3
Molecular Formula C11H7BrO3
Molecular Weight (g/mol) 267.08
MDL Number MFCD00094166
InChI Key HBUFBIFPUAHIDX-UHFFFAOYSA-N
PubChem CID 26419
IUPAC Name 4-bromo-3-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=C(O)C(Br)=C2C=CC=CC2=C1

Specifications

Melting Point 231°C
Quantity 5 g
Formula Weight 267.08
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-hydroxy-2-naphthoic Acid