missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-hydroxy-2-naphthoic Acid 97.0+%, TCI America™

Catalog No. b322925g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B322925G 25 g
B32295G 5 g
2 options

Catalog No. B322925G

Supplier: TCI America B322925G

Only null left

Chemical Identifiers

CAS 2208-15-3
Molecular Formula C11H7BrO3
Molecular Weight (g/mol) 267.08
MDL Number MFCD00094166
InChI Key HBUFBIFPUAHIDX-UHFFFAOYSA-N
PubChem CID 26419
IUPAC Name 4-bromo-3-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=C(O)C(Br)=C2C=CC=CC2=C1

Specifications

Melting Point 231°C
Quantity 25 g
Formula Weight 267.08
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-hydroxy-2-naphthoic Acid