missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chlorobenzotrifluoride, 98%

CAS: 402-04-0 | C7H3BrClF3 | 259.45 g/mol

$77.32 - $273.62

Chemical Identifiers

CAS 402-04-0
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00673985
InChI Key RVTIHGGJJMXISV-UHFFFAOYSA-N
Synonym 4-bromo-3-chlorobenzotrifluoride, 1-bromo-2-chloro-4-trifluoromethyl benzene, 1-bromo-2-chloro-4-trifluoromethyl-benzene, benzene, 1-bromo-2-chloro-4-trifluoromethyl, 3-chloro-4-bromobenzotrifluoride, 4-trifluoromethyl-2-chloro-bromo benzene, pubchem13478, ksc494s1t, 2-chloro-4-trifluoromethylbromobenzene, 4-cf3-2-chloro-bromo benzene
PubChem CID 14273266
IUPAC Name 1-bromo-2-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6197806
View Documents
Thermo Scientific Chemicals
H6197806
5 g
Each for $77.32
Only null left
 
AAH6197814
View Documents
Thermo Scientific Chemicals
H6197814
25 g
Each for $273.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

402-04-0
259.45
RVTIHGGJJMXISV-UHFFFAOYSA-N
14273266
C1=CC(=C(C=C1C(F)(F)F)Cl)Br
C7H3BrClF3
MFCD00673985
4-bromo-3-chlorobenzotrifluoride, 1-bromo-2-chloro-4-trifluoromethyl benzene, 1-bromo-2-chloro-4-trifluoromethyl-benzene, benzene, 1-bromo-2-chloro-4-trifluoromethyl, 3-chloro-4-bromobenzotrifluoride, 4-trifluoromethyl-2-chloro-bromo benzene, pubchem13478, ksc494s1t, 2-chloro-4-trifluoromethylbromobenzene, 4-cf3-2-chloro-bromo benzene
1-bromo-2-chloro-4-(trifluoromethyl)benzene

Specifications

402-04-0
MFCD00673985
4-bromo-3-chlorobenzotrifluoride, 1-bromo-2-chloro-4-trifluoromethyl benzene, 1-bromo-2-chloro-4-trifluoromethyl-benzene, benzene, 1-bromo-2-chloro-4-trifluoromethyl, 3-chloro-4-bromobenzotrifluoride, 4-trifluoromethyl-2-chloro-bromo benzene, pubchem13478, ksc494s1t, 2-chloro-4-trifluoromethylbromobenzene, 4-cf3-2-chloro-bromo benzene
C1=CC(=C(C=C1C(F)(F)F)Cl)Br
259.45
259.45
Liquid
C7H3BrClF3
5 g
RVTIHGGJJMXISV-UHFFFAOYSA-N
1-bromo-2-chloro-4-(trifluoromethyl)benzene
14273266
98%
4-Bromo-3-chlorobenzotrifluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only