accessibility menu, dialog, popup

UserName

FFFClBr

CAS RN 402-04-0

IUPAC Name: 1-bromo-2-chloro-4-(trifluoromethyl)benzene
Synonyms: 1-bromo-2-chloro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: RVTIHGGJJMXISV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results