missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylphenol 98.0+%, TCI America™

Catalog No. p-7119510
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21465G 5 g
B214625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21465G Supplier TCI America Supplier No. B21465G
Only null left

Chemical Identifiers

CAS 7463-51-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00002315
InChI Key WMUWDPLTTLJNPE-UHFFFAOYSA-N
Synonym 4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
PubChem CID 81970
IUPAC Name 4-bromo-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Br)C)O

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3,5-dimethylphenol