missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylphenol, 99%

CAS: 7463-51-6 | C8H9BrO | 201.063 g/mol

Supplier:  Thermo Scientific Chemicals B2168414

 View more versions of this product

Catalog No. AAB2168414


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-3,5-dimethylphenol
113°C to 117°C
C8H9BrO
25 g
4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
CC1=CC(=CC(=C1Br)C)O
201.063
201.07
7463-51-6
Phenol-like
MFCD00002315
2207127
WMUWDPLTTLJNPE-UHFFFAOYSA-N
4-bromo-3,5-dimethylphenol
81970
99%

Chemical Identifiers

7463-51-6
201.063
WMUWDPLTTLJNPE-UHFFFAOYSA-N
81970
CC1=CC(=CC(=C1Br)C)O
C8H9BrO
MFCD00002315
4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
4-bromo-3,5-dimethylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 231-255-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only