missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3,5-dimethoxybenzoic Acid 97.0+%, TCI America™
$32.72 - $88.56
Chemical Identifiers
| CAS | 56518-42-4 |
|---|---|
| Molecular Formula | C9H8BrO4 |
| Molecular Weight (g/mol) | 260.06 |
| MDL Number | MFCD01632140 |
| InChI Key | JNFZULSIYYVRJO-UHFFFAOYSA-M |
| Synonym | benzoic acid, 4-bromo-3,5-dimethoxy, zlchem 1334, acmc-1az7r, jnfzulsiyyvrjo-uhfffaoysa, 3,5-dimethoxy-4-bromobenzoic acid, 4-bromo-3,5-dimethoxybenzoic acid |
| PubChem CID | 2774026 |
| IUPAC Name | 4-bromo-3,5-dimethoxybenzoate |
| SMILES | COC1=CC(=CC(OC)=C1Br)C([O-])=O |
Chemical Identifiers
| 56518-42-4 | |
| 260.06 | |
| JNFZULSIYYVRJO-UHFFFAOYSA-M | |
| 2774026 | |
| COC1=CC(=CC(OC)=C1Br)C([O-])=O |
| C9H8BrO4 | |
| MFCD01632140 | |
| benzoic acid, 4-bromo-3,5-dimethoxy, zlchem 1334, acmc-1az7r, jnfzulsiyyvrjo-uhfffaoysa, 3,5-dimethoxy-4-bromobenzoic acid, 4-bromo-3,5-dimethoxybenzoic acid | |
| 4-bromo-3,5-dimethoxybenzoate |
Specifications
| 56518-42-4 | |
| White-Yellow | |
| MFCD01632140 | |
| benzoic acid, 4-bromo-3,5-dimethoxy, zlchem 1334, acmc-1az7r, jnfzulsiyyvrjo-uhfffaoysa, 3,5-dimethoxy-4-bromobenzoic acid, 4-bromo-3,5-dimethoxybenzoic acid | |
| COC1=CC(=CC(OC)=C1Br)C([O-])=O | |
| 260.06 | |
| 261.07 | |
| Crystalline Powder |
| 252°C | |
| C9H8BrO4 | |
| 5 g | |
| JNFZULSIYYVRJO-UHFFFAOYSA-M | |
| 4-bromo-3,5-dimethoxybenzoate | |
| 2774026 | |
| ≥97.0% (HPLC,T) | |
| 4-Bromo-3,5-dimethoxybenzoic Acid |
Safety and Handling
TSCA : No