missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethoxybenzoic acid, 98%

Catalog No. AAB2144906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2144906 5 g
AAB2144914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2144906 Supplier Thermo Scientific Chemicals Supplier No. B2144906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56518-42-4 | C9H8BrO4 | 260.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56518-42-4
Molecular Formula C9H8BrO4
Molecular Weight (g/mol) 260.06
MDL Number MFCD01632140
InChI Key JNFZULSIYYVRJO-UHFFFAOYSA-M
Synonym benzoic acid, 4-bromo-3,5-dimethoxy, zlchem 1334, acmc-1az7r, jnfzulsiyyvrjo-uhfffaoysa, 3,5-dimethoxy-4-bromobenzoic acid, 4-bromo-3,5-dimethoxybenzoic acid
PubChem CID 2774026
IUPAC Name 4-bromo-3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1Br)C([O-])=O

Specifications

Melting Point 254°C to 258°C
Quantity 5 g
Formula Weight 261.07
Percent Purity 98%
Chemical Name or Material 4-Bromo-3,5-dimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only