missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3,5-dihydroxybenzoic Acid 97.0+%, TCI America™
$104.38 - $521.94
Chemical Identifiers
| CAS | 16534-12-6 |
|---|---|
| Molecular Formula | C7H5BrO4 |
| Molecular Weight (g/mol) | 233.017 |
| MDL Number | MFCD00002513 |
| InChI Key | NUTRHYYFCDEALP-UHFFFAOYSA-N |
| Synonym | benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r |
| PubChem CID | 86023 |
| IUPAC Name | 4-bromo-3,5-dihydroxybenzoic acid |
| SMILES | C1=C(C=C(C(=C1O)Br)O)C(=O)O |
Chemical Identifiers
| 16534-12-6 | |
| 233.017 | |
| NUTRHYYFCDEALP-UHFFFAOYSA-N | |
| 86023 | |
| C1=C(C=C(C(=C1O)Br)O)C(=O)O |
| C7H5BrO4 | |
| MFCD00002513 | |
| benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r | |
| 4-bromo-3,5-dihydroxybenzoic acid |
Specifications
| 16534-12-6 | |
| C7H5BrO4 | |
| 25 g | |
| NUTRHYYFCDEALP-UHFFFAOYSA-N | |
| 4-bromo-3,5-dihydroxybenzoic acid | |
| 86023 | |
| ≥97.0% (HPLC,T) | |
| 4-Bromo-3,5-dihydroxybenzoic Acid |
| 273°C | |
| MFCD00002513 | |
| benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r | |
| C1=C(C=C(C(=C1O)Br)O)C(=O)O | |
| 233.017 | |
| 233.02 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-3066&(3)-1572
TSCA : Yes