missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals

$414.85 - $414.85

Chemical Identifiers

CAS 651027-00-8
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237.00
MDL Number MFCD18917156
InChI Key NLLRAEQVOZOSBB-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
PubChem CID 11424897
IUPAC Name 4-bromo-3,5-difluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(Br)C(F)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5990403
View Documents
Thermo Scientific Chemicals
H5990403
1 g
Each for $414.85
Only null left
 
Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

651027-00-8
237.00
NLLRAEQVOZOSBB-UHFFFAOYSA-N
11424897
OC(=O)C1=CC(F)=C(Br)C(F)=C1
C7H3BrF2O2
MFCD18917156
benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
4-bromo-3,5-difluorobenzoic acid

Specifications

651027-00-8
C7H3BrF2O2
1 g
benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
OC(=O)C1=CC(F)=C(Br)C(F)=C1
237.00
237
4-Bromo-3,5-difluorobenzoic acid
181°C to 185°C
MFCD18917156
9620931
NLLRAEQVOZOSBB-UHFFFAOYSA-N
4-bromo-3,5-difluorobenzoic acid
11424897
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only