accessibility menu, dialog, popup

UserName

CAS RN 651027-00-8

IUPAC Name: 4-bromo-3,5-difluorobenzoic acid
Synonyms: 4-bromo-3,5-difluorobenzoic acid
Molecular Weight (g/mol): 237
Molecular Formula: C7H3BrF2O2
InChi Key: NLLRAEQVOZOSBB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(Br)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results