missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAH5990403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5990403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5990403 Supplier Thermo Scientific Chemicals Supplier No. H5990403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 651027-00-8 | Molecular Formula: C7H3BrF2O2 | Molecular Weight: 237.00 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 651027-00-8
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237.00
MDL Number MFCD18917156
InChI Key NLLRAEQVOZOSBB-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
PubChem CID 11424897
IUPAC Name 4-bromo-3,5-difluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(Br)C(F)=C1

Specifications

Melting Point 181°C to 185°C
Quantity 1 g
Beilstein 9620931
Formula Weight 237
Percent Purity 96%
Chemical Name or Material 4-Bromo-3,5-difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only