missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-difluoroanisole 98.0+%, TCI America™

Catalog No. B339225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B339225G 25 g
B33925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B339225G Supplier TCI America Supplier No. B339225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 202865-61-0
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.02
MDL Number MFCD00143268
InChI Key GEJMNTXYFBBTFH-UHFFFAOYSA-N
Synonym 4-bromo-3,5-difluoroanisole, 3,5-difluoro-4-bromoanisole, 4-bromo-3,5-difluorophenyl methyl ether, benzene, 2-bromo-1,3-difluoro-5-methoxy, pubchem4090, acmc-1cp5s, ksc490k4f, 2,6-difluoro-4-methoxybromobenzene, 2,6-difluoro-4-methoxy-bromobenzene, 2-bromo-1,3-difluoro-5-methoxy-benzene
PubChem CID 2724984
IUPAC Name 2-bromo-1,3-difluoro-5-methoxybenzene
SMILES COC1=CC(F)=C(Br)C(F)=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 223.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-3,5-difluoroanisole