accessibility menu, dialog, popup

UserName

H3COFBrF

CAS RN 202865-61-0

IUPAC Name: 2-bromo-1,3-difluoro-5-methoxybenzene
Synonyms: 2-bromo-1,3-difluoro-5-methoxybenzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: GEJMNTXYFBBTFH-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(Br)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results