missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrophenylacetic Acid 97.0+%, TCI America™

Catalog No. p-7123993
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B51591G 1 g
B51595G 5 g
2 options

Catalog No. B51591G

Supplier: TCI America B51591G

Only null left

Chemical Identifiers

CAS 6127-11-3
Molecular Formula C8H5BrNO4
Molecular Weight (g/mol) 259.04
MDL Number MFCD05663950
InChI Key LBZPHZBNFDOCCR-UHFFFAOYSA-M
PubChem CID 271551
SMILES C1=CC(=C(C=C1Br)[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 167°C
Quantity 1 g
Synonym 2-4-bromo-2-nitrophenyl acetic acid, 4-bromo-2-nitrophenylacetic acid, 4-bromo-2-nitrophenyl acetic acid, 4-bromo-2-nitro-phenyl-acetic acid, benzeneacetic acid,4-bromo-2-nitro, benzeneacetic acid, 4-bromo-2-nitro, acmc-1bae0, 4-bromo-2-nitrophenylaceticacid, 4-bromo-2-nitro-phenylacetic acid, 2-4-bromo-2-nitro-phenyl acetic acid
Formula Weight 260.04
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitrophenylacetic Acid