accessibility menu, dialog, popup

UserName

OOBrNOO

CAS RN 6127-11-3

IUPAC Name: 2-(4-bromo-2-nitrophenyl)acetate
Synonyms: 2-(4-bromo-2-nitrophenyl)acetate
Molecular Weight (g/mol): 259.04
Molecular Formula: C8H5BrNO4
InChi Key: LBZPHZBNFDOCCR-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=C(Br)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results