missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitroanisole 98.0+%, TCI America™

Catalog No. p-7123298
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40105G 5 g
B401025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40105G Supplier TCI America Supplier No. B40105G
Only null left

Chemical Identifiers

CAS 33696-00-3
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00055529
InChI Key ORBHQHXVVMZIDP-UHFFFAOYSA-N
Synonym 4-bromo-2-nitroanisole, 2-methoxy-5-bromonitrobenzene, 4-brom-2-nitroanisole, 4-bromo-2-nitro anisole, 5-bromo-2-methoxynitrobenzene, 3-nitro-4-methoxy bromobenzene, 1-methoxy-2-nitro-4-bromobenzene, benzene, 4-bromo-1-methoxy-2-nitro, pubchem5341, 4-brom-2-nitroanisol
PubChem CID 118533
IUPAC Name 4-bromo-1-methoxy-2-nitrobenzene
SMILES COC1=C(C=C(C=C1)Br)[N+](=O)[O-]

Specifications

Melting Point 87°C
Quantity 5 g
Formula Weight 232.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-nitroanisole
TSCA No
Recommended Storage Refrigerator