accessibility menu, dialog, popup

UserName

CAS RN 33696-00-3

IUPAC Name: 4-bromo-1-methoxy-2-nitrobenzene
Synonyms: 4-bromo-1-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: ORBHQHXVVMZIDP-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)