missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitroanisole, 97%

Catalog No. p-7056119
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6487706 5 g
AAH6487714 25 g
AAH6487722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6487706 Supplier Thermo Scientific Chemicals Supplier No. H6487706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33696-00-3 | C7H6BrNO3 | 232.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33696-00-3
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD00055529
InChI Key ORBHQHXVVMZIDP-UHFFFAOYSA-N
Synonym 4-bromo-2-nitroanisole, 2-methoxy-5-bromonitrobenzene, 4-brom-2-nitroanisole, 4-bromo-2-nitro anisole, 5-bromo-2-methoxynitrobenzene, 3-nitro-4-methoxy bromobenzene, 1-methoxy-2-nitro-4-bromobenzene, benzene, 4-bromo-1-methoxy-2-nitro, pubchem5341, 4-brom-2-nitroanisol
PubChem CID 118533
IUPAC Name 4-bromo-1-methoxy-2-nitrobenzene
SMILES COC1=C(C=C(C=C1)Br)[N+](=O)[O-]

Specifications

Melting Point 85°C to 100°C
Quantity 5 g
Beilstein 2259903
Formula Weight 232.03
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-nitroanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-642-5
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only