missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzoic acid, 98+%

Catalog No. p-7043850
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0307106 5 g
AAL0307114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0307106 Supplier Thermo Scientific Chemicals Supplier No. L0307106
Only null left

CAS: 68837-59-2 | C8H7BrO2 | 215.046 g/mol

A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 68837-59-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00040905
InChI Key RVCJOGNLYVNRDN-UHFFFAOYSA-N
Synonym 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5
PubChem CID 99570
IUPAC Name 4-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Br)C(=O)O

Specifications

Melting Point 182°C to 184°C
Quantity 5 g
Beilstein 2249816
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-methylbenzoic acid
Hazard Category H301-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 272-437-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only