Learn More
4-Bromo-2-methylbenzoic acid, 97%
CAS: 68837-59-2 | C8H7BrO2 | 215.05 g/mol
$135.06 - $135.06
Chemical Identifiers
| CAS | 68837-59-2 |
|---|---|
| Molecular Formula | C8H7BrO2 |
| Molecular Weight (g/mol) | 215.05 |
| InChI Key | RVCJOGNLYVNRDN-UHFFFAOYSA-N |
| Synonym | 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5 |
| PubChem CID | 99570 |
| IUPAC Name | 4-bromo-2-methylbenzoic acid |
| SMILES | CC1=C(C=CC(=C1)Br)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC443400050
|
Thermo Scientific Chemicals
443400050 |
5 g |
Each for $135.06
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 68837-59-2 | |
| 215.05 | |
| 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5 | |
| 4-bromo-2-methylbenzoic acid |
| C8H7BrO2 | |
| RVCJOGNLYVNRDN-UHFFFAOYSA-N | |
| 99570 | |
| CC1=C(C=CC(=C1)Br)C(=O)O |
Specifications
| 68837-59-2 | |
| Authentic | |
| C8H7BrO2 | |
| 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5 | |
| CC1=C(C=CC(=C1)Br)C(=O)O | |
| 215.05 | |
| 215.05 | |
| 4-Bromo-2-methylbenzoic acid |
| 182.0°C to 184.0°C | |
| 96% min. (HPLC) | |
| 5 g | |
| RVCJOGNLYVNRDN-UHFFFAOYSA-N | |
| 4-bromo-2-methylbenzoic acid | |
| 99570 | |
| 97% |
Safety and Handling
GHS H Statement Toxic if swallowed. Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 272-437-7
RUO – Research Use Only