missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methyl-6-nitroaniline 98.0+%, TCI America™

Catalog No. B37301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37301G 1 g
B37305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37301G Supplier TCI America Supplier No. B37301G

Chemical Identifiers

CAS 77811-44-0
Molecular Formula C7H7BrN2O2
Molecular Weight (g/mol) 231.049
MDL Number MFCD00052919
InChI Key ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
Synonym 4-bromo-2-methyl-6-nitro-phenylamine, 4-bromo-2-methyl-6-nitrobenzenamine, 2-amino-5-bromo-3-nitrotoluene, 4-bromo-6-nitro-o-toluidine, benzenamine, 4-bromo-2-methyl-6-nitro, 4-bromo-2-methyl-6-nitrophenylamine, 4-bromo-2-methyl-6-nitro-aniline, pubchem3813, acmc-1bg8u, ksc496e7t
PubChem CID 522679
IUPAC Name 4-bromo-2-methyl-6-nitroaniline
SMILES CC1=CC(=CC(=C1N)[N+](=O)[O-])Br

Specifications

Melting Point 146°C
Quantity 1 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-methyl-6-nitroaniline