accessibility menu, dialog, popup

UserName

CAS RN 77811-44-0

IUPAC Name: 4-bromo-2-methyl-6-nitroaniline
Synonyms: 4-bromo-2-methyl-6-nitroaniline
Molecular Weight (g/mol): 231.05
Molecular Formula: C7H7BrN2O2
InChi Key: ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(=C1N)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)